Learn More
Invitrogen™ Dematin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114181
Description
Immunogen sequence: KKPPIYKQRE SVGGSPQTKH LIEDLIIESS KFPAAQPPDP NQPAKIETDY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 86%; Guinea Pig: 79%; Horse: 85%; Human: 100%; Mouse: 93%; Rat: 93%.
The protein encoded by this gene is an actin binding and bundling protein that plays a structural role in erythrocytes, by stabilizing and attaching the spectrin/actin cytoskeleton to the erythrocyte membrane in a phosphorylation-dependent manner. This protein contains a core domain in the N-terminus, and a headpiece domain in the C-terminus that binds F-actin. When purified from erythrocytes, this protein exists as a trimer composed of two 48 kDa polypeptides and a 52 kDa polypeptide. The different subunits arise from alternative splicing in the 3' coding region, where the headpiece domain is located. Disruption of this gene has been correlated with the autosomal dominant Marie Unna hereditary hypotrichosis disease, while loss of heterozygosity of this gene is thought to play a role in prostate cancer progression. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Dematin | |
Polyclonal | |
Unconjugated | |
DMTN | |
AI325486; dematin; dematin actin binding protein; dematin actin-binding protein; DMT; Dmtn; Epb4.9; Epb49; Erythrocyte membrane protein band 4.9; erythrocyte membrane protein band 4.9 (dematin); erythrocyte protein band 4.9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2039 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q08495 | |
DMTN | |
Immunogen sequence: KKPPIYKQRE SVGGSPQTKH LIEDLIIESS KFPAAQPPDP NQPAKIETDY (aa 142-191). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.