Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Demi Fraser Broth Base, Sold by AquaPhoenix Scientific Products for Science Education

Medium used for the selective enrichment of Listeria species including Listeria monocytogenes

$64.73 - $279.16

Specifications

For Use With (Application) Selective enrichment of Listeria species including Listeria monocytogenes
Product Type Broth Base
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Specimen Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Specimen Price Quantity & Availability  
S26822 Products for Science Education
View Documents
3M™
BP0118500
500 g
Each for $64.73
Add to cart
 
S26821 Products for Science Education
View Documents
3M™
BP0118005
2.5 kg
Each for $279.16
Add to cart
 
Description

Description

  • 3M™ dehydrated media
  • Convenience of making it when needed
Specifications

Specifications

Selective enrichment of Listeria species including Listeria monocytogenes
Broth Base
Safety and Handling

Safety and Handling

ShelfLife : 4 year

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit