Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Demonstration Balance for Student Lever Experiments Products for Science Education

This GSC International balance stand helps teachers with in-class demonstrations

Manufacturer:  GSC Go Science Crazy 415003

 View more versions of this product

Catalog No. S40853A


Add to cart

Requires :
  • Meter stick
  • Weights


Description

Description

  • For balance demonstration
  • Metal support and cast iron base supports knife edge clamp
  • Size: 3.75 x 3.75 x 6.5in.
  • Weight: 1.2 lb.
Specifications

Specifications

for Student Lever Experiments
Fulcrum Holder
544g
“U” shaped metal support and cast Iron base
1.2 lb.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit