Learn More
DemoSlide Products for Science Education
No need to bother with wet-mount slide preparation.
Specifications
Material Plastic
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S16441 Products for Science Education
View Documents
Optimize Technologies Inc.
GSMSA/2
2/Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Simply pipet or pour 5mL of live protozoa into a DemoSlide tube and view under your microscope
  • No spills, broken glass slides or evaporation — just easy-to-use tubes
  • Uses the Davstar Cen-Slide™
Two options:
  • Package of 120 DemoSlide tubes and caps — with this option, we recommend purchasing at least several sets of DemoSlide microscope stage adapters
  • Complete DemoSlide Kit: Includes 120 tubes and caps, stage adapter and a 12-place tube rack
Specifications
Specifications
Plastic