Learn More
Corning™ Denatured Collagen Dissolvable Microcarriers
Corning Dissolvable Microcarriers are designed to simplify your cell harvest, separation, and concentration processes. The microcarriers are sterile, and denatured collagen coated to enchance cell attachment, maximizing yield and viability. These microcarriers can be quickly dissolved, allowing easy collection of grown cells without the need for microcarrier separation.
Manufacturer:  Corning™ 4982
 View more versions of this product
Catalog No. 07-202-361
Add to cart
Description
Description
  • Denatured Collagen coated for enhanced cell attachment
  • Highest Sterility Assurance Level (SAL) of 10-6
  • Certified nonpyrogenic
  • Optimized for MSC applications
  • Bead size: Fully hydrated average size 200 to 300µm
  • 5000cm2/gram dry weight
Specifications
Specifications
200 to 300μm fully hydrated
Denatured Collagen Dissolvable Microcarriers.
Polygalacturonic acid (PGA) polymer chains cross linked via calcium ions; Denatured Collagen coated
Cell therapy, Vaccine production, Cell Scale-up, Cell Banking
5000cm2/gram dry weight
100g