Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Denatured salmon testes DNA, phenol extracted, Alfa Aesar™

Click to view available options
Quantity:
2 x 10mg
10mg
Quantity 2 x 10mg
Packaging Plastic bottle
Physical Form Liquid
Chemical Name or Material Denatured salmon testes DNA