Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dendra2 Rabbit anti-Tag, liquid, TrueRAB™

Rabbit Recombinant Monoclonal Antibody
Supplier: OriGene TA591007S

Description
Dendra2 is an improved version of a green-to-red photoswitchable fluorescent protein Dendra, derived from octocoral Dendronephthya species.Specifications
Dendra2 | |
Recombinant Monoclonal | |
Unconjugated | |
Dendra2 | |
Recombit protein of Dendra2 expressed in HEK293 cells | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Western Blot | |
OR-5C7 | |
Tissue Culture Supernatant with 1% BSA and 0.02% sodium azide | |
Rabbit | |
30 μL | |
Primary | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction