Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dendritic Cell Marker Rat anti-Mouse, Clone: AP-MAB0801, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
$177.16 - $493.00
Specifications
Antigen | Dendritic Cell Marker |
---|---|
Clone | AP-MAB0801 |
Dilution | Flow Cytometry 1:100-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:25-1:50, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBT85474100
|
Novus Biologicals
NB11085474100UG |
100 μg |
Each for $493.00
|
|
NB018042
|
Novus Biologicals
NB110854740.025MG |
0.025 mg |
Each for $177.16
|
|
Description
Dendritic Cell Marker Monoclonal specifically detects Dendritic Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
Dendritic Cell Marker | |
Flow Cytometry 1:100-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:25-1:50, CyTOF-ready | |
Monoclonal | |
Purified | |
Mouse | |
DCs, Dendritic cell | |
IgG2 | |
Protein G purified | |
This reacts with Dendritic Cell Inhibitory Receptor 2 on mouse dendritic cells. |
AP-MAB0801 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Rat | |
A 0.2 um filtered solution in PBS with No Preservative | |
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse dendritic cells (DCs). | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title