Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dendritic Cells Rat anti-Mouse, Clone: MIDC-8, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
Specifications
Antigen | Dendritic Cells |
---|---|
Clone | MIDC-8 |
Dilution | Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:25 |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10064608
|
Novus Biologicals
NB10064608 |
1 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB017619
|
Novus Biologicals
NB100646080.25ML |
0.25 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Dendritic Cells Monoclonal specifically detects Dendritic Cells in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Dendritic Cells | |
Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:25 | |
Monoclonal | |
Supernatant | |
RUO | |
DCs, Dendritic cell, FDC, FDCs, Follicular dendritic cell | |
IgG2a | |
Tissue culture supernatant | |
NB100-64608 is located in the cytoplasm as distinct spots near the nucleus. The antigen is found in dendritic cells; MIDC-8 does not stain thymic epithelial cells which are the major contaminants in thymic dendritic cell preparations. The antigen is also absent in blood cells, bone marrow cells or macrophages isolated from the peritoneal cavity. The antigen is found in interdigitating cells in T cell dependent areas of secondary lymphoid organs, the medulla of thymus, veiled cells, and very weakly in Langerhans cells. |
MIDC-8 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Rat | |
Mouse | |
Made to Mouse DENDRITIC/INTERDIGITATING CELLS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title