Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus 1 NS1 Mouse anti-Virus, Unconjugated, Clone: 912 , MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen Dengue Virus 1 NS1
Applications ELISA, Flow Cytometry
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF21901MI
View Documents
MilliporeSigma
MABF2190100UG
100 μg
Each for $372.00
Add to cart
 
MABF21902MI
View Documents
MilliporeSigma
MABF219025UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

Dengue Virus 1 NS1 Monoclonal antibody specifically detects Dengue Virus 1 NS1 in Virus samples. It is validated for ELISA, Flow Cytometry.
Specifications

Specifications

Dengue Virus 1 NS1
Unconjugated
Mouse
DENV NS1 protein
Monoclonal
Protein G Purified
ELISA, Flow Cytometry
Purified
Virus
Purified recombinant Dengue virus 1 NS1 protein expressed in mammalian cells.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit