Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue virus 4 NS1 Mouse anti-Viral, Clone: 971314, Novus Biologicals™

Mouse MonoclonalAntibody

Specifications

Antigen Dengue virus 4 NS1
Clone 971314
Dilution Western Blot 1 ug/mL
Applications Western Blot
Format Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
MAB9444SP View Documents Novus BiologicalsSupplier Diversity Partner
MAB9444SP
25 ug Unconjugated Each for $102.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Dengue virus 4 NS1 Monoclonal antibody specifically detects Dengue virus 4 NS1 in Viral samples. It is validated for Western Blot.
Specifications

Specifications

Dengue virus 4 NS1
Western Blot 1 ug/mL
Purified
Virus
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
IgG2b
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
971314
Western Blot
Mouse
RUO
Human embryonic kidney cell line HEK293-derived recombinant Dengue Virus 4 NS1 protein Asp1-Ala352 Accession # NP_740318
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Detects recombinant Dengue Virus 4 NS1 in direct and capture ELISAs and Western blots. In direct ELISAs and Western Blots, no cross-reactivity with recombinant Dengue Virus 1, 2 and 3 NS1 proteins was observed.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit