Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue virus NS1 Mouse anti-Virus, Clone: HM027, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13533
Description
Mouse monoclonal antibody raised against dengue virus NS1.
- High specificity
- High reactivity
Specifications
Dengue virus NS1 | |
Monoclonal | |
Unconjugated | |
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
Mouse | |
1 mg | |
Primary | |
Virus | |
Liquid |
ELISA, Immunofluorescence, Lateral Flow | |
HM027 | |
Mouse monoclonal antibody raised against dengue virus NS1. | |
In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
Protein A purification | |
RUO | |
This antibody reacts with NS1 of dengue virus. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction