Learn More
Dengue virus NS1 Mouse anti-Virus, Clone: HM100, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13535
Catalog No. 89-033-498
Add to cart
Description
Description
Mouse monoclonal antibody raised against dengue virus NS1.
  • High specificity
  • High reactivity
Specifications
Specifications
Dengue virus NS1
HM100
Mouse monoclonal antibody raised against dengue virus NS1.
In 10 mM PBS, pH 7.4.
Mouse
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Lateral Flow
Unconjugated
Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
Native purified dengue virus antigen.
RUO
Primary
Virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)