Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dengue Virus Type 1-4 NS1 Monoclonal Antibody (5168)
GREENER_CHOICE

Catalog No. PIMA533248
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
PIMA533248 1 mL
1 options

Catalog No. PIMA533248

Supplier: Invitrogen™ MA533248

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dengue Virus Type 1-4 NS1 Monoclonal Antibody for ICC/IF, ELISA, LF

Dengue fever and dengue hemorrhagic fever are acute febrile diseases, found in the tropics and Africa. Caused by one of four closely related virus serotypes of the genus Flavivirus, family Flaviviridae, each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur. Dengue is transmitted to humans by the Aedes aegypti (rarely Aedes albopictus) mosquito, which feeds during the day.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus Type 1-4 NS1
Applications ELISA, Lateral Flow, Immunocytochemistry
Classification Monoclonal
Clone 5168
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Dengue NS1; Dengue pan
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.