Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dengue Virus Type 2 Envelope (Domain III) (strain New Guinea C/PUO-218 hybrid) Monoclonal Antibody (201)
GREENER_CHOICE

Catalog No. PIMA530000
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA530000 100 μL
1 options

Catalog No. PIMA530000

Supplier: Invitrogen™ MA530000

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E/Envelope Protein (Domain III).

Dengue fever and dengue hemorrhagic fever are acute febrile diseases, found in the tropics and Africa. Caused by one of four closely related virus serotypes of the genus Flavivirus, family Flaviviridae, each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur. Dengue is transmitted to humans by the Aedes aegypti (rarely Aedes albopictus) mosquito, which feeds during the day.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus Type 2 Envelope (Domain III) (strain New Guinea C/PUO-218 hybrid)
Applications Western Blot
Classification Monoclonal
Clone 201
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Dengue 2 envelope; Dengue envelope; envelope protein
Host Species Mouse
Immunogen Recombinant Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E/Envelope Protein (Domain III) Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.