Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DenLine Protection Plus Ladies Full Cut A-Line Design with Traditional Collar, 39 in. (99.1 cm) Long Lab Coat- Fluid Resistant (AAMI Level 2)
Click to view available options
Color:
Ceil Blue
Green
Royal Blue
White
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
Primary Applications
· Anatomic Pathology · Clinical Pathology · Clinical Laboratory · Phlebotomy · Specimen Collection · Blood Collection, Testing, & Manufacturing · Human & Animal Research · Academia · Dialysis · Dental · Clean Room ISO 5/Class 100 - Medical Device, Pharma, Electronics
Wearer Protection Provided
- Full Cut Design provides superior top of leg protection when seated.
- Assists wearers comply with OSHA 29 CFR 1910
- AAMI PB70 Level 2 Protection against Blood & OPIM; Front, Sleeves, Shoulders and Back
Wearer Comfort
- Overall Air Permeability 18 CFM (ASTM D737)
- Comfortable when worn in room temperatures ± 72°F (22.2°C) with controlled humidity <50%.
Other Features
- Stain Resistant Fabric and Cuffs (blood, bleach and all aqueous reagents)
- Anti-static Fabric prevents false instrument readings, damage to computers
- Low Lint Fabric prevents sample contamination, ideal for molecular testing
- Durable Construction
- 3 Pockets – Double Stitched & Bar Tacked No Failure design
- Stainless Snap Closures – Lifetime Guaranteed
- Durable Cuffs for Overgloving – Bleach Safe all colors
- Back Belt Detail
- Hang Loop, Outside Back of Collar
- Side Vents for pant pocket access
- Sizes run large…XXS to 5XL, Available White, Ceil Blue, Royal Blue and Green.
- 225 Commercial Wash Tested; Low Lifetime Cost per Wash.
- Autoclavable Fabric (4 Min @ 274°F - 134.4°C)
- Launder using Home/Laundromat or Commercial Equipment; Instructions furnished meet CDC/HICPAC Recommendations for processing medical bloodborne Linen
Specifications
Type | Lab Coat |
Size | X-Small |
Gender | Women's |
Material | Textured Polyester, Carbon |
Color | Green |
Closure Type | Snaps |
Cuff Style | Knit |
Disposable | Reusable |
Sterility | Non-sterile |
Resistance | Hydrostatic Pressure >32.5 cm (AATCC 127) |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction