Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine Protection Plus Unisex 41 in. (104 cm) Long Lab Coat- Fluid Resistant Barrier (AAMI Level 2)
SDP

Catalog Number 22412028
Click to view available options
Color:
Black
Ceil Blue
Emerald Green
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Style protects wearer against incidental splash of Blood, Body Fluids, Aerosols, etc.; AAMI Level 2 Barrier; Assists with OSHA compliance; Air Permeability 18 CFM for Comfort; Stain resistant to blood, bleach, and ALL aqueous reagents; Anti-static: Low Lint; Durable; 225 Wash Tested.

Primary Applications

· Anatomic Pathology · Clinical Pathology · Clinical Laboratory · Phlebotomy · Specimen Collection · Blood Collection, Testing, & Manufacturing · Human & Animal Research · Academia · Dialysis · Dental · Clean Room ISO 5/Class 100 - Medical Device, Pharma, Electronics

Wearer Protection Provided

  • Style Offers DenLine's Highest Barrier Protection against strike through from incidental splash of Blood and OPIM…tested Hydrostatic Pressure >32.5 CM (AATCC 127) and Impact Penetration <1.0 grams (AATCC 42) after completing 225 Hot Water Commercial Wash Cycles. Assists wearers comply with OSHA 29 CFR 1910
  • 360 Degree protection Front as well as Shoulders and Back. All Fabric used is fluid resistant…tested AAMI PB70 Level 2 Barrier. No Cotton Fabric used on Shoulders or Back as it absorbs…would not meet OSHA 29 if splashed with blood or OPIM traveling rearward either side of the employee's head.

Wearer Comfort

  • DenLine UltraLite fabric with high air permeability (35 CFM) is used to make the Back of Coat for wearer comfort. Overall Air Permeability of Coat Front and Back is 18 CFM (ASTM D737).
  • Coat Comfortable when worn in normal room temperatures ± 72°F (22.2°C) with controlled humidity <50%.

Other Features

  • Stain Resistant Fabric and Cuffs (blood, bleach and ALL aqueous reagents & liquids)
  • Anti-static Fabric prevents false instrument readings, damage to computers
  • Low Lint Fabric prevents sample contamination, ideal for molecular testing
  • Durable Construction
    • 3 Pockets – Double Stitched & Bar Tacked No Failure design
    • Stainless Snap Closures – Lifetime Guaranteed
    • Durable Cuffs for Overgloving – Bleach Safe all colors
  • Hang Loop, Outside Back of Collar
  • Side Vents for pant pocket access
  • Choice of 5 Colors White, Ceil Blue, Royal, Black and Green, Sizes XS to 5XL
  • Autoclavable Fabric (4 Min @ 274°F - 134.4°C)
  • 225 Commercial Wash Tested; Low Lifetime Cost per Wash
  • Launder using Home/Laundromat or Commercial Equipment; Instructions furnished meet CDC/HICPAC Recommendations for processing medical bloodborne Linen
Type Long Lab Coat
Size 3X-Large
Gender Unisex
Material Polyester, Carbon
Color Emerald Green
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Hydrostatic Pressure >32.5 cm (AATCC 127)
No. of Pockets 3
Description Durable Knit Cuffs for Overgloving - Bleach Safe all Colors
Collar Type Traditional
Finish Fluid Resitant, Anti-Static
Latex Free Yes
Size (English) Chest 54 to 56 in.
Size (English) Hip 50 to 53.5 in.
Size (Metric) Chest 137 to 142 cm
Size (Metric) Hip 127 to 136 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection
For Use With (Application) Protection Against Incidental Splash of Blood & OPIM
Length (English) 41 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 44 in.
Length (Metric) 104 cm
Length (Metric) Cuff 10.1 cm
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit