Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Protection Plus™ Fluid-Resistant Short-Length Lab Jackets

Fluid-resistant lab jackets assist in OSHA compliance

Manufacturer:  DENLINE UNIFORMS INC DL147MWHT

 View more versions of this product

Catalog No. 22-412-163


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

OSHA
White
3
Unisex
4 in.
Round
38 to 39 in.
31.5 in.
Snap
Spun Woven Polyester
Medium
EZ Snap
10.16cm
38 to 40 in.
80cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit