Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DENND1A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | DENND1A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
DENND1A Polyclonal specifically detects DENND1A in Human samples. It is validated for Western Blot.Specifications
DENND1A | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
57706 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Connecdenn, Connecdenn 1, DENN/MADD domain containing 1A, FAM31ADENN domain-containing protein 1A, FLJ21129, FLJ38464, KIAA1608connecdenn, Protein FAM31A, RP11-230L22.3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 270-527 of human DENND1A (NP_079096.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title