Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DENND4C Mouse anti-Human, Mouse, Rat, Clone: 300000000, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00 - $603.50
Specifications
Antigen | DENND4C |
---|---|
Clone | 300000000 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
Description
DENND4C Monoclonal specifically detects DENND4C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
DENND4C | |
1 mg/ml | |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
bA513M16.3, C9orf55B, DENN/MADD domain containing 4C, DKFZp686I09113, FLJ20686 | |
Full Length Human Recombinant DENND4C protein (NM_017925) | |
Primary | |
Store at -20°C. |
300000000 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.4), 50% Glycerol | |
55667 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title