Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DENND4C Mouse anti-Human, Mouse, Rat, Clone: 300000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP31104025UG

 View more versions of this product

Catalog No. NB120200


Add to cart

Description

Description

DENND4C Monoclonal specifically detects DENND4C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

DENND4C
300000000
Unconjugated
PBS (pH 7.4), 50% Glycerol
bA513M16.3, C9orf55B, DENN/MADD domain containing 4C, DKFZp686I09113, FLJ20686
Full Length Human Recombinant DENND4C protein (NM_017925)
Protein G purified
RUO
Primary
55667
Western Blot, Immunofluorescence, Immunoprecipitation
1 mg/ml
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Purified
Mouse
IgG1
25 μg
Store at -20°C.
Monoclonal
Human, Mouse, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit