Thermo Scientific™ Dense Storage Options Available on GSA/VA Contract for Federal Government customers only.

Double storage capacity and cut storage costs by 50%. Dense Storage Options feature standard Cryoboxes that work with standard freezer racks.

Manufacturer: Thermo Scientific™ 369640

 View more versions of this product

Catalog No. 12-570-218

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Vial Type Cryobank Tubes
    For Use With Thermo Scientific Nunc Cryobank and Bank-It Vial Systems
    Color white
    Description Cardboard Cryo box with 13 x 13 divider
    No. per Pack 24
    No. per Case 24
    Capacity 169 Cryobank Tubes
    Item Description Nunc cardboard box

    Double storage capacity and cut storage costs by 50%. The Thermo Scientific™ Dense Storage Options use standard Thermo Scientific™ CryoBox™ sizes that work with a lab's existing freezer racks. By using standard boxes with 13 x 13 tube separators, (169) 1mL tubes fit in a standard size CryoBox.

    • Available in economical cardboard or long lasting plastic cryobox options
    • Fits (169) 1mL Cryobank tubes for the most dense storage available

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit