Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Density Samples Sets Collection
Each set of 12 is made from the same material in different lengths.
$24.00
Specifications
For Use With (Application) | To determine the mass and volume of each sample to determine the density |
---|
Includes:
- Four sets: aluminum, brass, PVC and polypropylene
- Wooden holder with linseed coating for water protection
- Instructions
Catalog Number | Mfr. No. | Product Type | Includes | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Includes | Material | Price | Quantity & Availability | |||
S68357
![]() |
Science First![]() 6112045 |
Density sample set | Wooden holder with linseed coating for Water protection, instructions | Tekaform |
Each for $24.00
|
|
Description
- Determine the mass and volume of each sample to determine the density
- Sets are available separately or as the complete collection
Specifications
To determine the mass and volume of each sample to determine the density |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title