Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ deoC (Escherichia coli) Recombinant Protein

Used for SDS-PAGE

Manufacturer:  Abnova™ P4996

Catalog No. 89-941-793


Add to cart

Description

Description

Sequence: MGSSHHHHHHSSGLVPRGSHMTDLKASSLRALKLMDLTTLNDDDTDEKVIALCHQAKTPVGNTAAICIYPRFIPIARKTLKEQGTPEIRIATVTNFPHGNDDIDIALAETRAAIAYGADEVDVVFPYRALMAGNEQVGFDLVKACKEACAAANVLLKVIIETGELKDEALIRKASEISIKAGADFIKTSTGKVAVNATPESARIMMEVIRDMGVEKTVGFKPAGGVRTAEDAQKYLAIADELFGADWADARHYRFGASSLLASLLKALGHGDGKSASSY
Specifications

Specifications

NP_418798
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
29.9kDa
Escherichia coli expression system
>95% by SDS-PAGE
100 ug
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
ECK4373|JW4344|dra|thyR|tlr
deoC
Escherichia coli
1 mg/mL
Liquid
948902
deoC (Escherichia coli) Recombinant Protein
Conventional Chromatography
Loading 3 ug protein in 15% SDS-PAGE
MGSSHHHHHHSSGLVPRGSHMTDLKASSLRALKLMDLTTLNDDDTDEKVIALCHQAKTPVGNTAAICIYPRFIPIARKTLKEQGTPEIRIATVTNFPHGNDDIDIALAETRAAIAYGADEVDVVFPYRALMAGNEQVGFDLVKACKEACAAANVLLKVIIETGELKDEALIRKASEISIKAGADFIKTSTGKVAVNATPESARIMMEVIRDMGVEKTVGFKPAGGVRTAEDAQKYLAIADELFGADWADARHYRFGASSLLASLLKALGHGDGKSASSY
RUO
deoC
Escherichia coli expression system
His
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit