Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxycholic acid, 98.5-101%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
83-44-3
C24H40O4
392.58
MFCD00003673
KXGVEGMKQFWNSR-LLQZFEROSA-N
deoxycholic acid,deoxycholate,desoxycholic acid,choleic acid,cholerebic,cholorebic,degalol,deoxycholatic acid,droxolan,pyrochol
222528
CHEBI:28834
(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
CC(CCC(=O)O)C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C
83-44-3
C24H40O4
392.58
MFCD00003673
KXGVEGMKQFWNSR-LLQZFEROSA-N
deoxycholic acid,deoxycholate,desoxycholic acid,choleic acid,cholerebic,cholorebic,degalol,deoxycholatic acid,droxolan,pyrochol
222528
CHEBI:28834
(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
CC(CCC(=O)O)C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C
Infrared Spectrum Authentic
Melting Point 172°C to 178°C
Color Pink to White
Loss on Drying 1 % max. (140°C, 4 hrs) (under vacuum < 5 mmHg)
Quantity 100g
Specific Rotation +53° to +57° (20°C, 589nm) (c=1, EtOH)
Packaging Glass bottle
Ignition Residue 0.2 % max.
Assay Percent Range 98.5%
Beilstein 10, IV, 1608
Merck Index 15, 2901
Formula Weight 392.57
Physical Form Powder
Percent Purity 98.0 to 102.0% (on dried basis) (Acid)
Chemical Name or Material Deoxycholic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning