Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Deoxyribonuclease I (Bovine)

Used for Enzyme

Manufacturer:  Abnova™ P5307

Catalog No. 89-941-797


Add to cart

Description

Description

#REF!
Specifications

Specifications

Native protein chromatographically purified from Bovine Pancreas
Enzyme Activity
282217
Deoxyribonuclease I (Bovine)
25000 Units
RUO
DNASE1
≥2,000 Kunitz units per mg dry weight. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0.
None
P00639
Lyophilized
31.3kDa
Native protein chromatographically purified from Bovine Pancreas
Store at 4°C on dry atmosphere.
DNASE1
Deoxyribonuclease I Assay; Nick translation; DNA mapping; Isolation of nuclear RNA and protein; Plasmid construction; RNA polymerase synthesis of RNA probes and RT-PCR.
Bovine
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit