Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Deoxyribonuclease I (Bovine)
Used for Enzyme
Supplier: Abnova™ P5221
Description
#REF!Specifications
P00639 | |
282217 | |
Deoxyribonuclease I (Bovine) | |
5 mg | |
RUO | |
DNASE1 | |
≥2,000 Kunitz units per mg dry weight. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0. | |
None | |
Lyophilized |
Enzyme Activity | |
31.3kDa | |
Native protein chromatographically purified from Bovine Pancreas | |
Store at 4°C on dry atmosphere. | |
DNASE1 | |
Deoxyribonuclease I Assay; DNA removal in primary cell isolation to decreases viscosity providing better yields; DNA removal in bioprocessing applications. | |
Bovine | |
Native protein chromatographically purified from Bovine Pancreas |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction