Learn More
Abnova™ Deoxyribonuclease I (Bovine), Molecular Biology Grade
Used for Enzyme
Manufacturer:  Abnova™ P5308
Catalog No. 89-941-798
Add to cart
Description
Description
#REF!
Specifications
Specifications
P00639
Lyophilized
31.3kDa
Native protein chromatographically purified from Bovine Pancreas
Store at 4°C on dry atmosphere. Each vial contains approximately 2 mg glycine and 2 umoles calcium per 10,000 units of DNase I. Reconstituted the entire vial in 5 ml provides the equivalent of a 1 mg/ml solution.
DNASE1
Deoxyribonuclease I Assay; Nick translation; DNA mapping; Isolation of nuclear RNA and protein; Plasmid construction; RNA polymerase synthesis of RNA probes and RT-PCR.
≥2,000 Kunitz units per mg dry weight. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0.
None
Enzyme Activity
282217
Deoxyribonuclease I (Bovine), Molecular Biology Grade
2500 Units
RUO
DNASE1
Native protein chromatographically purified from Bovine Pancreas
Bovine