Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Deoxyribonuclease II (Porcine)
Used for Enzyme
Supplier: Abnova™ P5297
Description
#REF!Specifications
Enzyme Activity | |
Deoxyribonuclease II (Porcine) | |
20000 Units | |
RUO | |
Deoxyribonuclease II Assay | |
Pig | |
Native protein chromatographically purified from Porcine Spleen by a modification of the procedure of Bernardi et al., Biochim. Biophys. Acta, 129, 1 (1966) |
38kDa | |
Native protein chromatographically purified from Porcine Spleen by a modification of the procedure of Bernardi et al., Biochim. Biophys. Acta, 129, 1 (1966) | |
Store at -20°C. | |
DEOXYRIBONUCLEASE | |
≥12,000 units per mg protein. One Unit causes an increase in absorbance at 260 nm of 0.001 per minute at 25°C, pH 4.6 using highly polymerized DNA as substrate. | |
None | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction