Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonucleic Acid Sodium Salt, Fisher BioReagents

Click to view available options
Quantity:
250mg
1g
5g
10g
Packaging:
Amber Glass
68938-01-2
C17H15BrFNO3
380.213
GXZCYECDYOBPGH-OAHLLOKOSA-N
73904662
(2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
C1=CC(=CC(=C1)Br)CC(C(=O)O)NC(=O)CC2=CC=C(C=C2)F
68938-01-2
C17H15BrFNO3
380.213
GXZCYECDYOBPGH-OAHLLOKOSA-N
73904662
(2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
C1=CC(=CC(=C1)Br)CC(C(=O)O)NC(=O)CC2=CC=C(C=C2)F
Deoxyribonucleic Acid Sodium Salt is used in several biochemical research applications.
Quantity 10g
Packaging Amber Glass
Identification Pass Test
Loss on Drying 14% max.
Color White
Formula Weight 6000000
Physical Form Solid
Chemical Name or Material Deoxyribonucleic Acid Sodium Salt, From Salmon Sperm

Recommended Storage : 4°C