Learn More
DEP-1/CD148 Mouse anti-Human, Alexa Fluor™ 532, Clone: MEM-CD148/05, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221790AF532
Catalog No. NP221790A32
Add to cart
Description
Description
DEP-1/CD148 Monoclonal antibody specifically detects DEP-1/CD148 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
DEP-1/CD148
MEM-CD148/05
Flow Cytometry
CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody MEM-CD148/05 recognizes CD148, a highly glycosylated up to 250 kDa receptor-like protein tyrosin phosphatase expressed mainly in lymphocytes, myeloid cells and epithelial cells.
Flow Cytometry
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
PTPRJ
Human recombinant CD148 (amino acids 1-444).
0.1 mL
Primary
5795
Human