Learn More
BioVision IncSupplier Diversity Partner Deproteinizing Sample Preparation Kit II: Deproteinizing Sample Preparation Kit II: Rapid & Convenient, Non-PCA method to deproteinize a variety of samples. 200 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K823200
Deproteinizing Sample Preparation Kit II: The study of biofluids, cell, bacteria, yeast, and tissue lysates has delivered vital information regarding their biochemical composition. These biological samples comprise a rich, complex list of lipids, proteins, small organic molecules, ions, and, depending on the sample, exogenous molecules (i.e. drugs). BioVision's Deproteinizing Sample Preparation Kit II offers an excellent alternative when metabolite lability affects the accurate detection and estimation of small molecule concentrations and when organic solvents or ultrafiltration cannot be used. TCA precipitates proteins by lowering the sample pH drastically. After removal of precipitated proteins, the pH of the sample is neutralized with neutralization buffer that is provided in the kit. The kit is easy to follow, convenient, and can be utilized for the preparation of a large number of samples in parallel and can be directly used in various bioassays.
Catalog No. 50-111-9938