Learn More
Apexbio Technology LLC Esomeprazole Magnesium 161973-10-0 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC B22005.1
Esomeprazole Magnesium is a small-molecule inhibitor targeting the H /K -ATPase enzyme on gastric parietal cells It is designed to inhibit this proton pump thereby reducing gastric acid secretion Esomeprazole Magnesium exerts its biological activity primarily through selective and irreversible binding to H /K -ATPase inhibiting the final step in gastric acid secretion Based on these pharmacological properties Esomeprazole Magnesium holds research potential in studies of gastric acidity regulation gastroesophageal reflux disease (GERD) inflammation-related mucosal damage and investigations of pharmacokinetics and pharmacodynamics of proton pump inhibitors
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.