Learn More
Sklar InstrumentsSupplier Diversity Partner TC Derf Needle Holder, Sklar, 4-3/4 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218004
TC Derf Needle Holder, Sklar, 4-3/4 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. The Sklar TC Derf Needle Holder features short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures. Frequently utilized in ophthalmic, dental, or plastic procedures, the shorter length allows the Derf Needle Holder to be easily controlled in smaller surgical areas. Tungsten Carbide instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips and a length of 4-3/4 inches.
Catalog No. 50-143-7694