Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DERL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549629

Catalog No. PA549629

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Functional component of endoplasmic reticulum-associated degradation (ERAD) for misfolded lumenal glycoproteins, but not that of misfolded nonglycoproteins. May act by forming a channel that allows the retrotranslocation of misfolded glycoproteins into the cytosol where they are ubiquitinated and degraded by the proteasome. May mediate the interaction between VCP and the degradation substrate. In contrast to DERL1, it is not involved in the degradation of MHC class I heavy chains following infection by cytomegaloviruses. May play a role in cell proliferation.
Specifications

Specifications

DERL2
Polyclonal
PBS with 0.09% sodium azide
Q9GZP9
Derlin-2, Degradation in endoplasmic reticulum protein 2, DERtrin-2, Der1-like protein 2, F-LAN-1, F-LANa, DERL2, DER2, FLANA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51009
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DERL2
Liquid
DERL2
KLH conjugated synthetic peptide between 191-218 amino acids from the C-terminal region of human DERL2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit