Derma Sciences First Aid Adhesive Bandages

Manufacturer: DUKAL CORPORATION 1304000

 View more versions of this product

Catalog No. 19-065749

  Check Availability
Add to Cart

    Specifications

    Color Natural
    For Use With (Application) For prevent skin maceration
    Latex Free Yes
    Size (English) 0.75 x 3in.
    Size (Metric) 1.9 x 7.5cm
    Sterile Yes
    Product Type Sheer Bandage
    Quantity 1500 Pack

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit