Learn More
Alfa Aesar™ Dermenkephalin
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66523LB0
 View more versions of this product
Catalog No. AAJ66523LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
955.15
5mg
Tyr-D-Met-Phe-His-Leu-Met-Asp-NH2
-30° to -10°C
HPLC
Solid
RUO
Dermenkephalin