Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DES Mouse anti-Human, Clone: OTI4G1, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502328
Description
Desmin (approximately 53 kDa) exhibits a high degree of tissue specificity, its expression being predomitly confined to all types of muscle cells (cardiac, skeletal and smooth muscle). Regulation of desmin expression is stage and tissue-specific, since it is induced during terminal development of, for example, skeletal muscle cell differentiation. In skeletal en cardiac muscle cells desmin is localized in the Z-disk region and at the intercalated disk. The expression pattern of desmin in smooth muscle is much more heterogenous. Coexpression of vimentin and desmin has been observed in tumors derived from muscle tissue, i.e. rhabdomyosarcomas and leiomyosarcomas. Furthermore, during myocard dysfunction dramatic changes in the distribution of desmin have been observed.Specifications
DES | |
Monoclonal | |
0.63 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P17661 | |
CDCD3, CMD1F, CSM1, CSM2, LGMD1D, LGMD1E, LGMD2R | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
1674 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4G1 | |
Unconjugated | |
DES | |
Liquid | |
DES | |
Human fragment corresponding to amino acids 104-391 of human DES produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title