Learn More
Tocris Bioscience™ [des-octanoyl]-Ghrelin (rat)
Major circulating form of ghrelin; devoid of activity at GHS receptor
Manufacturer:  Tocris Bioscience™ 2951/1
Catalog No. 29-511-R
Add to cart
This item is not returnable. View return policy
Description
Description
Non-acylated, major circulating isoform of ghrelin that does not bind to the ghrelin receptor (GHS-R1a), nor induce growth hormone release.
Specifications
Specifications
[des-octanoyl]-Ghrelin (rat)
1mg
[Des-octanoyl]-Ghrelin (rat)
3188.63
Gly-Ser-Ser-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg