Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desert Hedgehog/Dhh Rat anti-Mouse, Clone: RM0086-6A12, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP121726
Description
Desert Hedgehog/Dhh Monoclonal antibody specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot.Specifications
Desert Hedgehog/Dhh | |
RM0086-6A12 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, HHG-3MGC35145 | |
Rat | |
IgG2 | |
0.1 mg | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of dhh N-terminal fragment. | |
Mouse |
Western Blot | |
LYOPH | |
Western Blot 1:500-1:1000 | |
Purified | |
DHH | |
Mouse recombinant dhh N-terminal fragment | |
Protein G purified | |
RUO | |
Primary | |
50846 | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only