Learn More
Desert Hedgehog/Dhh Rat anti-Mouse, DyLight 488, Clone: RM0087-7C8, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP121727G
Catalog No. NBP121727G
Add to cart
Description
Description
Desert Hedgehog/Dhh Monoclonal antibody specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Desert Hedgehog/Dhh
RM0087-7C8
Western Blot
Purified
DHH
Mouse recombinant dhh C-terminal fragment
Protein G purified
Store at 4C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of dhh C-terminal fragment.
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, HHG-3MGC35145
Rat
IgG2
0.1 ml
Primary
50846
Mouse