Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desert Hedgehog/Dhh Rat anti-Mouse, DyLight 550, Clone: RM0086-6A12, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP121726R
Description
Desert Hedgehog/Dhh Monoclonal antibody specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot.Specifications
Desert Hedgehog/Dhh | |
RM0086-6A12 | |
Western Blot | |
Purified | |
DHH | |
Mouse recombinant dhh N-terminal fragment | |
Protein G purified | |
Store at 4C in the dark. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of dhh N-terminal fragment. |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, HHG-3MGC35145 | |
Rat | |
IgG2 | |
0.1 ml | |
Primary | |
50846 | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title