Learn More
Fisherbrand™ Desk Height Class 100 Clean Room/ESD Chair with Medium Back Products for Science Education
Fisherbrand Desk Height Class 100 Clean Room/ESD Chair with Medium Back features electro-static dissipating components for highly sensitive applications. Fully adjustable for standard 30 in high desks.
$398.00 - $504.00
Specifications
Adjustable (Height) 18 to 23 in
Base Style CHROME
Capacity 300 lbs
Seating Form LOW
Seating Height DESK HEIGHT
View More Specs
Products 8
Catalog Number Color With Armrest With Tilt Arm Style Price Quantity & Availability  
Catalog Number Color With Armrest With Tilt Arm Style Price Quantity & Availability  
S39949 Products for Science Education View Documents Blue NO NO Not adjustable Each for $398.00
Add to cart
S41151 Products for Science Education View Documents Blue YES NO Adjustable Each for $476.00
Add to cart
S39951 Products for Science Education View Documents Blue YES YES Adjustable Each for $504.00
Add to cart
S39952 Products for Science Education View Documents Black NO NO Not adjustable Each for $398.00
Add to cart
S39953 Products for Science Education View Documents Black YES NO Adjustable Each for $476.00
Add to cart
S39954 Products for Science Education View Documents Black YES YES Adjustable Each for $504.00
Add to cart
S39955 Products for Science Education View Documents Black NO YES Not adjustable Each for $425.00
Add to cart
S39956 Products for Science Education View Documents Blue NO YES Not adjustable Each for $425.00
Add to cart
Description
Description
  • Features ergonomic adjustments 
  • For 30 in high work surface
  • Larger Back Pad for extra comfort
  • Optional 6-way adjustable arms for proper ergonomic arm placement
  • Optional 8-degree forward, 3-degreee backward locking seat tilt
  • Suitable for Class 100 (ISO Class 5) Clean Rooms
  • Electro-Static Dissipating Vinyl Upholstered Seat and Back
  • Electro-Static Dissipating, Dual-Wheel, Non-Marring Casters for proper grounding
Certifications
ANSI/BIFMA, ANSI/ESD-STM-12.1-2006
Warranty and Services
15-year on parts, 5-year on upholstery, Lifetime on the pneumatic lift
Specifications
Specifications
18 to 23 in
300 lbs
DESK HEIGHT
YES
CHROME
LOW
YES