Learn More
ILEX LIFE SCIENCES LLC Deslorelin (GnRH agonist), Synthetic Peptide, 100 mg
SDP
Supplier:  ILEX LIFE SCIENCES LLC A2240100
Encompass_Preferred
Deslorelin is a synthetic nonapeptide analogue of the natural gonadotropin releasing hormone (GnRH) with potential antineoplastic activity. Deslorelin binds to and activates pituitary gonadotropin releasing hormone (GnRH) receptors. Deslorelin has a molecular weight of 1281.64 Dalton. MOLECULAR FORMULA: C64H83N17O12. AMINO ACID SEQUENCE: pGlu-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-NHC2H5. CAS REGISTRY NO.: 57773-65-6. PURITY: Greater than 99.0%. FORMULATION: Sterile-filtered white lyophilized (freeze-dried) powder. The Deslorelin peptide was lyophilized with no additives. OTHER NAMES: Gonadotropin-releasing hormone (GnRH) superagonist, luteinizing hormone-releasing hormone agonist, luteinising hormone-releasing hormone agonist, GnRH agonist, LHRH agonist.
Catalog No. 50-233-9484
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.