Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmethyl-YM 298198, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1177767-57-5
C17H21ClN4OS
364.892
SWKZWXJQIDXGLX-UHFFFAOYSA-N
desmethyl-ym 298198,6-amino-n-cyclohexyl-3-methylthiazolo 3,2-a benzimidazole-2-carboxamide hydrochloride,desmethyl ym 298198 hydrochloride
16759180
7-amino-N-cyclohexyl-1-methyl-[1,3]thiazolo[3,2-a]benzimidazole-2-carboxamide;hydrochloride
CC1=C(SC2=NC3=C(N12)C=C(C=C3)N)C(=O)NC4CCCCC4.Cl
This item is not returnable. View return policy
1177767-57-5
C17H21ClN4OS
364.892
SWKZWXJQIDXGLX-UHFFFAOYSA-N
desmethyl-ym 298198,6-amino-n-cyclohexyl-3-methylthiazolo 3,2-a benzimidazole-2-carboxamide hydrochloride,desmethyl ym 298198 hydrochloride
16759180
7-amino-N-cyclohexyl-1-methyl-[1,3]thiazolo[3,2-a]benzimidazole-2-carboxamide;hydrochloride
CC1=C(SC2=NC3=C(N12)C=C(C=C3)N)C(=O)NC4CCCCC4.Cl

Derivative of mGlu1 antagonist YM 298198 (Cat. No. 2448)

Derivative of the mGlu 1 -selective antagonist YM 298198 (Cat. No. 2448).
Quantity 50mg
Formula Weight 364.89
Percent Purity >98%
Chemical Name or Material Desmethyl-YM 298198

Recommended Storage : Desiccate at Room Temperature