Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Mouse anti-Human, PE/Atto594, Clone: OTI4G1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270565PEATT594

Catalog No. N270565P594


Add to cart

Description

Description

Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Desmin
OTI4G1
Flow Cytometry
Affinity Purified, Conjugated
DES
Recombinant protein fragment corresponding to amino acids 104-391 of human Desmin (NP_001918) produced in E.coli.
Immunogen affinity purified
Cytoskeleton Markers, Phospho Specific
Primary
1674.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit