Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Rabbit anti-Human, Mouse, Sheep, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB120152000.1ML

Catalog No. NBW1520001

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Desmin
1.0 mg/mL
Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:75-1:200, Immunohistochemistry-Frozen 1:10-1:500
P17661
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
Rabbit
IgG
Immunogen affinity purified
Cytoskeleton Markers
Primary
1674
Human, Mouse, Sheep
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.6) and 1.0% BSA with 0.1% Sodium Azide
Affinity Purified
DES
Synthetic peptide mapping near the C-terminus of human desmin.
53 kDa
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Desmin
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit