Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Rabbit anti-Human, Rat, Chicken, Hamster, Clone: DES/2960R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30764010UG

 View more versions of this product

Catalog No. NB122753


Add to cart

Description

Description

Desmin Monoclonal specifically detects Desmin in Human, Rat, Chicken, Hamster samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Desmin
DES/2960R
Unconjugated
10 mM PBS with 0.05% BSA
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
Recombinant human full-length Desmin protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human, Rat, Chicken, Hamster
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Rabbit
IgG
100 μg
Cell Biology, Cellular Markers, Phospho Specific
Primary
1674
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit