Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Rabbit anti-Human, Rat, Chicken, Hamster, Clone: DES/2960R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Desmin
Clone DES/2960R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122753
View Documents
Novus Biologicals
NBP30764010UG
100 μg
Each for $406.52
Add to cart
 
NB122754
View Documents
Novus Biologicals
NBP30764020UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Desmin Monoclonal antibody specifically detects Desmin in Human, Rat, Chicken, Hamster samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Desmin
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human, Rat, Chicken, Hamster
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
Recombinant human full-length Desmin protein
Monoclonal
Protein A or G purified
DES/2960R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cell Biology, Cellular Markers, Phospho Specific
10 mM PBS with 0.05% BSA
1674
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit