Learn More
Desmocollin-2 Mouse anti-Human, Mouse, Rat, Alexa Fluor™ 532, Clone: 7G6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP254519AF532
Catalog No. NP2419A32
Add to cart
Description
Description
Desmocollin-2 Monoclonal antibody specifically detects Desmocollin-2 in Human,Mouse,Rat samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
Desmocollin-2
7G6
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
ARVD11, Cadherin family member 2, CDHF2DGII/III, desmocollin 2, desmocollin-2, Desmocollin-3, Desmosomal glycoprotein II, desmosomal glycoprotein II/III, Desmosomal glycoprotein III, DG2, DSC3DKFZp686I11137
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
DSC2
Human desmocollin-2 extracellular domain (exact sequence is proprietary)
0.1 mL
Primary
1824